Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554427
Description
Immunogen sequence: AGARKKELLE RLAQQQQQLY APPPPAEQEE QALSPSPTNG NPLLPFPGAQ VRQNVKNRLL VKNQDVSISN VQPKT Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 69%.
RBM33 gene ontology annotations related to this gene include RNA binding.
Specifications
RBM33 | |
Polyclonal | |
Unconjugated | |
Rbm33 | |
3200001K10Rik; 6430512A10Rik; AA386890; DKFZp434D1319; DKFZp686F102; hypothetical protein MGC20460; MGC20460; proline rich 8; proline-rich protein 8; Prr8; Rbm33; RNA binding motif protein 33; RNA-binding motif protein 33; RNA-binding protein 33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
155435 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EV2 | |
Rbm33 | |
Recombinant protein corresponding to Human RBM33. Recombinant protein control fragment (Product #RP-92827). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction