Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553964 Supplier Invitrogen™ Supplier No. PA553964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPSAQVKPIV SASPPSRAVA GSRSSQGKTE VKVKPASPVA QPKEEAKTET EFPDEDEETR LYRLKIEEQK RLREEILKQK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

RBM33 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM33
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbm33
Gene Accession No. Q96EV2
Gene Alias 3200001K10Rik; 6430512A10Rik; AA386890; DKFZp434D1319; DKFZp686F102; hypothetical protein MGC20460; MGC20460; proline rich 8; proline-rich protein 8; Prr8; Rbm33; RNA binding motif protein 33; RNA-binding motif protein 33; RNA-binding protein 33
Gene Symbols Rbm33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM33. Recombinant protein control fragment (Product #RP-92826).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.