Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM34 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128735AF594
Description
RBM34 Polyclonal antibody specifically detects RBM34 in Human samples. It is validated for Western BlotSpecifications
RBM34 | |
Polyclonal | |
Western Blot | |
KIAA0117RNA-binding motif protein 34, RNA binding motif protein 34, RNA-binding protein 34 | |
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human RNA binding motif protein 34 using the numbering given in entry CAI21936.1 (GeneID 23029). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23029 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction