Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM34 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297638F
Description
RBM34 Polyclonal specifically detects RBM34 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RBM34 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| KIAA0117RNA-binding motif protein 34, RNA binding motif protein 34, RNA-binding protein 34 | |
| Produced in rabbits immunized with E. coli-derived Human RBM34 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 23029 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction