Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541855 Supplier Invitrogen™ Supplier No. PA541855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SKPKQGLNFT SKTAEGHPKS LFIGEKAVLL KTKKKGQKKS GRPKKQRKQK Sequence homology: Dog: 86%; Horse: 86%; Human: 100%; Pig: 86%; Rat: 76%.

RNA binding motif protein 34 (RBM34) is an RRM (RNA recognition motif)-containing protein. Alternative names for RBM34 Antibody include KIAA0117 antibody, RNA-binding protein 34 antibody, RNA-binding motif protein 34 antibody, RNA binding motif protein 34 antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM34
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBM34
Gene Accession No. P42696
Gene Alias 4930547K05Rik; 6330444B11; D8Ertd233e; KIAA0117; Rbm34; RGD1310161; RNA binding motif protein 34; RNA-binding motif protein 34; RNA-binding protein 34
Gene Symbols RBM34
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RBM34 (aa 381-430).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23029
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.