Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587666 Supplier Invitrogen™ Supplier No. PA587666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSSLEPQIQP VYVPVPKQTI KKTKRNEEEE STSQIERPLS QEPAKKVKAK KKHTNAEKKL ADRESALASA DLEEEIHQKQ GQKRKNSQPG VKVADRKILD DTEDTVVSQR KKIQINQEEE RLKNERTVFV GNLPVTCNKK KLKSFFKEYG QIESVRFRSL IPAEGTLSKK LAAIKRKIHP DQKNINAYVV FKEESAATQA LKRNGAQIAD GFRIRVDLAS E; Positive Samples: HeLa, Jurkat, A-549, 293T, K-562; Cellular Location: Nucleus, nucleolus.

RNA binding motif protein 34 (RBM34) is an RRM (RNA recognition motif)-containing protein. Alternative names for RBM34 Antibody include KIAA0117 antibody, RNA-binding protein 34 antibody, RNA-binding motif protein 34 antibody, RNA binding motif protein 34 antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM34
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM34
Gene Accession No. P42696
Gene Alias 4930547K05Rik; 6330444B11; D8Ertd233e; KIAA0117; Rbm34; RGD1310161; RNA binding motif protein 34; RNA-binding motif protein 34; RNA-binding protein 34
Gene Symbols RBM34
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-280 of human RBM34 (NP_0558292).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23029
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.