Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599701
Description
Antibody detects endogenous levels of total RBM34.
RNA binding motif protein 34 (RBM34) is an RRM (RNA recognition motif)-containing protein. Alternative names for RBM34 Antibody include KIAA0117 antibody, RNA-binding protein 34 antibody, RNA-binding motif protein 34 antibody, RNA binding motif protein 34 antibody.
Specifications
RBM34 | |
Polyclonal | |
Unconjugated | |
RBM34 | |
4930547K05Rik; 6330444B11; D8Ertd233e; KIAA0117; Rbm34; RGD1310161; RNA binding motif protein 34; RNA-binding motif protein 34; RNA-binding protein 34 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23029, 52202 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P42696, Q8C5L7 | |
RBM34 | |
A synthesized peptide derived from human RBM34(Accession P42696), corresponding to amino acid residues A344-A394. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction