Learn More
RBM34 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302293A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RNA binding motif protein 34 (RBM34) is an RRM (RNA recognition motif)-containing protein. Alternative names for RBM34 Antibody include KIAA0117 antibody, RNA-binding protein 34 antibody, RNA-binding motif protein 34 antibody, RNA binding motif protein 34 antibody.Specifications
RBM34 | |
Polyclonal | |
Unconjugated | |
RBM34 | |
RNA-binding protein 34, RNA-binding motif protein 34, KIAA0117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23029 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P42696 | |
RBM34 | |
Between 75 and 125 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.