Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107233 Supplier Invitrogen™ Supplier No. PA5107233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

RBM38 is a probable RNA-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM38
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene RBM38
Gene Accession No. Q9H0Z9
Gene Alias CLL-associated antigen KW-5; dJ800J21.2; fi25e12; HSRNASEB; hypothetical protein; RBM38; RNA binding motif protein 38; rna-binding; RNA-binding motif protein 38; RNA-binding protein 38; RNA-binding region (RNP1, RRM) containing 1; RNA-binding region containing 1; RNA-binding region-containing protein 1; RNPC1; rnpc1 protein; Seb4; seb4 mRNA; SEB4B; SEB4D; Seb4l; ssDNA-binding protein SEB4; wu:fi25e12; YF-55; zgc:92336
Gene Symbols RBM38
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RBM38 (aa 189-239).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55544
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.