Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM38 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7682R

Description
RBM38 is a probable RNA-binding protein.Specifications
RBM38 | |
Polyclonal | |
Unconjugated | |
RBM38 | |
BOS_13172; CLL-associated antigen KW-5; dJ800J21.2; fi25e12; HSRNASEB; hypothetical protein; Rbm38; RNA binding motif protein 38; RNA-binding motif protein 38; RNA-binding protein 38; RNA-binding region (RNP1, RRM) containing 1; RNA-binding region containing 1; RNA-binding region-containing protein 1; Rnpc1; rnpc1 protein; SEB4; seb4 mRNA; SEB4B; SEB4D; Seb4l; ssDNA-binding protein SEB4; wu:fi25e12; YF-55; zgc:92336 | |
Rabbit | |
Protein A | |
RUO | |
55544 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9H0Z9 | |
RBM38 | |
KLH conjugated synthetic peptide derived from human RBM38. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction