Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM39 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993605
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3605 10 μL
50-199-3604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3605 Supplier Active Motif Supplier No. 61670
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is an RNA binding protein and possible splicing factor. The encoded protein is found in the nucleus, where it colocalizes with core spliceosomal proteins. Studies of a mouse protein with high sequence similarity to this protein suggest that this protein may act as a transcriptional coactivator for JUN/AP-1 and estrogen receptors. Multiple transcript variants encoding different isoforms have been observed for this gene.

Specifications

Antigen RBM39
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene RBM39
Gene Accession No. Q14498
Gene Alias 1500012C14Rik; 2310040E03Rik; B330012G18Rik; C79248; Caper; CAPER alpha; CAPERalpha; Coactivator of activating protein 1 and estrogen receptors; coactivator of activating protein-1 and estrogen receptors; Coactivator of AP-1 and ERs; FSAP59; functional spliceosome-associated protein 59; HCC1; Hepatocellular carcinoma protein 1; R75070; RBM39; RNA binding motif protein 39; RNA-binding motif protein 39; RNA-binding protein 39; RNA-binding region (RNP1, RRM) containing 2; RNA-binding region-containing 2; RNA-binding region-containing protein 2; Rnp1; Rnpc2; Rrm; Splicing factor HCC1; Transcription coactivator CAPER
Gene Symbols RBM39
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human RBM39.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9584
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.