Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24667125UL
Description
RBM4 Polyclonal antibody specifically detects RBM4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RBM4 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
E9PM61 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5936 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
DKFZp547K0918, FLJ36727, hLark, Lark homolog, MGC75138, RBM4. LARK, RBM4ALARK, RNA binding motif protein 4, RNA-binding motif protein 4, RNA-binding motif protein 4a, RNA-binding protein 4, transcriptional coactivator CoAZ, ZCCHC21, ZCRB3A, zinc finger CCHC-type and RNA binding motif 3A | |
This antibody was developed against Recombinant Protein corresponding to amino acids: YGKVLECDIIKNYGFVHIEDKTAAEDAIRNLHHYKLHGVNINVEASKNK | |
25 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction