Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM4 Mouse anti-Human, Clone: 3H3F12, Proteintech
SDP

Catalog No. 501736387 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6387 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6387 Supplier Proteintech Group Inc Supplier No. 602921IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May play a role in alternative splice site selection during pre-mRNA processing.

Specifications

Antigen RBM4
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3H3F12
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM4
Gene Accession No. Q9BWF3
Gene Alias hLark, LARK, Lark homolog, RBM4, RBM4A, RNA binding motif protein 4, RNA binding motif protein 4a, RNA binding protein 4, ZCCHC21, ZCRB3A
Gene Symbols RBM4
Host Species Mouse
Immunogen RBM4 Fusion Protein Ag2190
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5936
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.