Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587843 Supplier Invitrogen™ Supplier No. PA587843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVKLFIGNLP REATEQEIRS LFEQYGKVLE CDIIKNYGFV HIEDKTAAED AIRNLHHYKL HGVNINVEAS KNKSKTSTKL HVGNISPTCT NKELRAKFEE YGPVIECDIV KDYAFVHMER AEDAVEAIRG LDNTEFQGKR; Positive Samples: LO2, K562, mouse eye, mouse brain, rat brain; Cellular Location: Cytoplasm, Cytoplasmic granule, Nucleus, Nucleus speckle, nucleolus.

RNA-binding factor involved in multiple aspects of cellular processes like alternative splicing of pre-mRNA and translation regulation. Modulates alternative 5'-splice site and exon selection. Acts as a muscle cell differentiation-promoting factor. Activates exon skipping of the PTB pre-mRNA during muscle cell differentiation. Antagonizes the activity of the splicing factor PTBP1 to modulate muscle cell-specific exon selection of alpha tropomyosin. Binds to intronic pyrimidine-rich sequence of the TPM1 and MAPT pre-mRNAs. Required for the translational activation of PER1 mRNA in response to circadian clock. Binds directly to the 3'-UTR of the PER1 mRNA. Exerts a suppressive activity on Cap-dependent translation via binding to CU-rich responsive elements within the 3'UTR of mRNAs, a process increased under stress conditions or during myocytes differentiation. Recruits EIF4A1 to stimulate IRES-dependent translation initiation in respons to cellular stress. Associates to internal ribosome entry segment (IRES) in target mRNA species under stress conditions. Plays a role for miRNA-guided RNA cleavage and translation suppression by promoting association of AGO2-containing miRNPs with their cognate target mRNAs. Associates with miRNAs during muscle cell differentiation. Binds preferentially to 5'-CGCGCG[GCA]-3' motif in vitro. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM4
Gene Accession No. Q8C7Q4, Q9BWF3
Gene Alias 4921506I22Rik; hLark; LARK; lark homolog; Lark1; Mlark; RBM4; RBM4A; RNA binding motif protein 4; rna-binding; RNA-binding motif protein 4; RNA-binding motif protein 4a; RNA-binding protein 4; transcriptional coactivator CoAZ; Unknown (protein for MGC:128953); ZCCHC21; ZCRB3A; zinc finger CCHC-type and RNA binding motif 3A
Gene Symbols RBM4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human RBM4 (NP_0028872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19653, 293663, 5936
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.