Learn More
Invitrogen™ RBM42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113715
Description
Immunogen sequence: DPSDYVRAMR EMNGKYVGSR PIKLRKSMWK DRNLDVVRKK QKEKKKLGLR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Binds (via the RRM domain) to the 3'-untranslated region (UTR) of CDKN1A mRNA.
Specifications
| RBM42 | |
| Polyclonal | |
| Unconjugated | |
| Rbm42 | |
| 1700003D06Rik; 3100004P22Rik; MGC10433; RBM42; RGD1306184; RNA binding motif protein 42; RNA-binding motif protein 42; RNA-binding protein 42 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79171 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BTD8 | |
| Rbm42 | |
| Synthetic peptide directed towards the C terminal region of human RBM42 (aa 431-480). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.