Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560460 Supplier Invitrogen™ Supplier No. PA560460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MALEVPEPLG EDKKKGKPEK LKRCIRTAAG SSWEDPSLLE WDADDFRIFC GDLGNEVNDD ILARAFSRFP SFLKAKVIRD KRTGKTKGYG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Binds (via the RRM domain) to the 3'-untranslated region (UTR) of CDKN1A mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM42
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbm42
Gene Accession No. Q91V81, Q9BTD8
Gene Alias 1700003D06Rik; 3100004P22Rik; MGC10433; RBM42; RGD1306184; RNA binding motif protein 42; RNA-binding motif protein 42; RNA-binding protein 42
Gene Symbols Rbm42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBM42. Recombinant protein control fragment (Product #RP-100383).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68035, 79171
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.