Learn More
RBM42 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305139AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Binds (via the RRM domain) to the 3′-untranslated region (UTR) of CDKN1A mRNA.Specifications
RBM42 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q91V81, Q9BTD8 | |
RBM42 | |
Between 100 and 150 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RBM42 | |
MGC10433, RNA-binding motif protein 42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
68035, 79171 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.