Learn More
RBM42 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305138A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish and Bovine
Binds (via the RRM domain) to the 3′-untranslated region (UTR) of CDKN1A mRNA.Specifications
RBM42 | |
Polyclonal | |
Unconjugated | |
RBM42 | |
MGC10433, RNA-binding motif protein 42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
68035, 79171 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q91V81, Q9BTD8 | |
RBM42 | |
Between 430 and 480 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.