Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM46 Antibody (OTI1D6), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273833MFV500
Description
RBM46 Monoclonal antibody specifically detects RBM46 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
RBM46 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
166863 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1D6 | |
Flow Cytometry | |
Cancer/testis antigen 68, CT68MGC27016, probable RNA-binding protein 46, RNA binding motif protein 46, RNA-binding motif protein 46 | |
Full length human recombinant protein of human RBM46(NP_659416) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction