Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144778
Description
Immunogen sequence: QFTLLHLDYN FHRSSINSLS PVSATLSSGT PSVLPYTSRP YSYPGYPLSP TISLANGSHV GQRLCISNQ Antigen sequence identity: Mouse 90%; Rat 94%.
RBM46 is a protein with RNA binding function.
Specifications
RBM46 | |
Polyclonal | |
Unconjugated | |
RBM46 | |
cancer/testis antigen 68; CT68; EG633285; ENSMUSG00000033882; fe11d09; Gm761; LOW QUALITY PROTEIN: probable RNA-binding protein 46; probable RNA-binding protein 46; RBM46; RGD1560205; RNA binding motif protein 46; RNA-binding motif protein 46; wu:fe11d09; zgc:153695 | |
Rabbit | |
Affinity chromatography | |
RUO | |
166863 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TBY0 | |
RBM46 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 461-529. Recombinant protein control fragment (Product #RP-109767). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction