Learn More
Invitrogen™ RBM48 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113719
Description
Immunogen sequence: FLQTNPTGNE IMIGPLLPDI SKVDMHDDSL NTTANLIRHK LKEVISSVPK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 93%; Zebrafish: 83%.
RBM48 gene ontology annotations related to this gene include RNA binding.
Specifications
| RBM48 | |
| Polyclonal | |
| Unconjugated | |
| RBM48 | |
| AW548102; C030048B08Rik; C14H7orf64; C4H7orf64; C7orf64; HSPC304; MGC16142; Rbm48; RGD1310794; RNA binding motif protein 48; RNA-binding protein 48; UPF0712 protein C7orf64; UPF0712 protein C7orf64 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84060 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5RL73 | |
| RBM48 | |
| Synthetic peptide directed towards the C terminal region of human DKFZP564O0523. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.