Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541915 Supplier Invitrogen™ Supplier No. PA541915
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AAAATTSSYY GRDRSPLRRA AAMLPTVGEG YGYGPESELS QASAATRNSL Sequence homology: Cow: 100%; Dog: 93%; Goat: 85%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 93%; Rat: 86%.

Required for the translational activation of PER1 mRNA in response to circadian clock. Binds directly to the 3'-UTR of the PER1 mRNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBM4B
Gene Accession No. Q9BQ04
Gene Alias 4921506I22Rik; AI504630; AI506404; Lark2; Neosin; RBM30; RBM4B; RBM4L; RNA binding motif protein 4B; rna-binding; RNA-binding motif protein 30; RNA-binding motif protein 4B; RNA-binding protein 30; RNA-binding protein 4B; similar to zinc responsive protein ZD7; ZCCHC15; ZCCHC21B; ZCRB3B; zinc finger CCHC-type and RNA binding motif 3B; zinc responsive protein ZD7; zinc-responsive protein ZD7
Gene Symbols RBM4B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RBM4B (aa 290-339).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83759
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.