Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM5 Polyclonal Antibody

Catalog No. PIPA535963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535963 Supplier Invitrogen™ Supplier No. PA535963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: K562, THP-1. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (80%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DEF-3 and LUCA15 belong to an evolutionarily conserved family of RNA binding proteins and share similar expression patterns. Both DEF-3 and LUCA15 are highly expressed in adult heart and thymus as well as fetal kidney. Conversely, fetal thymus and adult kidney express very little DEF-3 and LUCA15. In the haemopoietic system of mice, the expression of DEF-3 is downregulated upon differentiation of progenitor cells into granulocytes but persists during macrophage development. Both DEF-3 and LUCA15 contain two zinc finger motifs, a bipartite nuclear signal and two RNA binding motifs. DEF-3 and LUCA15 are capable of specifically binding poly(G) RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RBM5
Gene Accession No. P52756
Gene Alias D030069N10Rik; G15; H37; LUCA15; Protein G15; Putative tumor suppressor LUCA15; Rbm5; Renal carcinoma antigen NY-REN-9; RMB5; RNA binding motif protein 5; RNA-binding motif protein 5; RNA-binding protein 5
Gene Symbols RBM5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RBM5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.