Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBM6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587864 Supplier Invitrogen™ Supplier No. PA587864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLEYVSSLDF WYCKRCKANI GGHRSSCSFC KNPREVTEAK QELITYPQPQ KTSIPAPLEK QPNQPLRPAD KEPEPRKREE GQESRLGHQK REAERYLPPS RREGPTFRRD RERESWSGET RQDGESKTIM LKRIYRSTPP EVIVEVLEPY VRLTTANVRI IKNRTGPMGH TYGFIDLDSH AEALRVVKIL QNLDPPFSID GKMVAVNLAT GKRRNDSGDH SDHMHYYQGK KYFRDRRGGG R; Positive Samples: A-549, 293T, HeLa, HepG2; Cellular Location: Nucleus.

DEF-3 and LUCA15 belong to an evolutionarily conserved family of RNA binding proteins and share similar expression patterns. Both DEF-3 and LUCA15 are highly expressed in adult heart and thymus as well as fetal kidney. Conversely, fetal thymus and adult kidney express very little DEF-3 and LUCA15. In the haemopoietic system of mice, the expression of DEF-3 is downregulated upon differentiation of progenitor cells into granulocytes but persists during macrophage development. Both DEF-3 and LUCA15 contain two zinc finger motifs, a bipartite nuclear signal and two RNA binding motifs. DEF-3 and LUCA15 are capable of specifically binding poly(G) RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM6
Gene Accession No. P78332
Gene Alias 3G2; 4930506F14Rik; DEF3; DEF-3; ETK2; g16; HEK2; HLC-11; Lung cancer antigen NY-LU-12; lung cancer protooncogene 11; mKIAA4015; NY-LU-12; Protein G16; Rbm6; RGD1560367; RNA binding motif protein 6; RNA-binding motif protein 6; RNA-binding protein 6; RNA-binding protein DEF-3; TYRO6
Gene Symbols RBM6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 530-770 of human RBM6 (NP_0057681).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10180
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.