Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157429
Description
RBM7 Polyclonal specifically detects RBM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RBM7 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ11153, RNA binding motif protein 7, RNA-binding motif protein 7, RNA-binding protein 7 | |
Rabbit | |
Affinity purified | |
RUO | |
10179 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q9Y580 | |
RBM7 | |
Synthetic peptides corresponding to RBM7(RNA binding motif protein 7) The peptide sequence was selected from the middle region of RBM7. Peptide sequence SFNQSSSSQWRQGTPSSQRKVRMNSYPYLADRHYSREQRYTDHGSDHHYR. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Mouse: 92%; Rat: 92%; Guinea pig: 85%; Bovine: 78%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction