Learn More
RBM8a Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301033A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, X. tropicalis and Bovine
This gene encodes a protein with a conserved RNA-binding motif. The protein is found predomitly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.Specifications
| RBM8a | |
| Polyclonal | |
| Unconjugated | |
| RBM8A | |
| RNA binding motif protein 8A, Y14, RBM8, ZNRP, RBM8B, ZRNP1, BOV-1A, BOV-1B, BOV-1C, MDS014 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60365, 9939 | |
| 4° C | |
| Liquid |
| Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9CWZ3, Q9Y5S9 | |
| RBM8A | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.