Learn More
RBM8A/Y14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
Specifications
Specifications
| Antigen | RBM8A/Y14 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.22 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RBM8A |
| Gene Accession No. | Q27W01, Q9CWZ3, Q9Y5S9 |
| Gene Alias | Binder of OVCA1 1, BOV 1, BOV 1A, BOV 1B, BOV 1C, MDS014, RBM8, RBM8A, RBM8B, Ribonucleoprotein RBM8A, RNA binding motif protein 8A, RNA binding protein 8A, RNA binding protein Y14, Y14, ZNRP, ZRNP1 |
| Gene Symbols | Rbm8a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.