Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBMS1 Antibody (OTI2H1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273834AF700
Description
RBMS1 Monoclonal specifically detects RBMS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RBMS1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RBMS1 | |
Full length recombinant protein of human RBMS1 (NP_058520) produced in E.coli. | |
0.1 mL | |
5937.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2H1 | |
Western Blot, Immunohistochemistry | |
chromosome 2 open reading frame 12, DKFZp564H0764, HCC-4, MGC15146, MGC3331, MGC97258, MGC97270, MGC97282, MGC99543, MSSP1, MSSP-1, MSSP-2, MSSP-3, RNA binding motif, single stranded interacting protein 1, SCR2MGC70597, single-stranded-interacting protein 1, suppressor of cdc 2 (cdc13) with RNA binding motif 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction