Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541760 Supplier Invitrogen™ Supplier No. PA541760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TYMPATSAMQ GAYLPQYAHM QTTAVPVEEA SGQQQVAVET SNDHSPYTFQ Sequence homology: Cow: 93%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBMS1
Gene Accession No. P29558
Gene Alias 2600014B10Rik; AI255215; C2orf12; cervical cancer oncogene 4; c-myc gene single strand binding protein 2; EMBL:AAH87094.1}; HCC-4; hypothetical protein LOC526135; MSSP; MSSP1; MSSP-1; MSSP-2; MSSP-3; Rbms1; rbms1 {ECO:0000312; RNA binding motif single stranded interacting protein 1; RNA binding motif, single stranded interacting protein 1; RNA-binding motif, single-stranded-interacting protein 1; SCR2; Single-stranded DNA-binding protein MSSP-1; suppressor of cdc 2 (cdc13) with RNA binding motif 2; suppressor of CDC2 with RNA-binding motif 2; YC1
Gene Symbols RBMS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RBMS1 (aa 354-403).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5937
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.