Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBMS2 Antibody (PCRP-RBMS2-1B6), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314163APC
This item is not returnable.
View return policy
Description
RBMS2 Monoclonal antibody specifically detects RBMS2 in Human samples. It is validated for Flow CytometrySpecifications
RBMS2 | |
Monoclonal | |
APC | |
FLJ39093, FLJ40023, RNA binding motif, single stranded interacting protein 2, RNA-binding motif, single-stranded-interacting protein 2, SCR3FLJ43262, Suppressor of CDC2 with RNA-binding motif 3 | |
Recombinant full-length human RBMS2 protein (Uniprot: Q15434) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-RBMS2-1B6 | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
5939 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction