Learn More
Invitrogen™ RBMS2 Monoclonal Antibody (3B12)

Description
Peptide Sequence: HHHSYLMQPS GSVLTPGMDH PISLQPASMM GPLTQQLGHL SLSSTGTYMP TAAAMQGAYI SQYTPVPSSS VSVEESSGQQ NQVAVDAPSE HGVYSFQFN*.
Specifications
Specifications
| Antigen | RBMS2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3B12 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Rbms2 |
| Gene Accession No. | Q15434 |
| Gene Alias | RBMS2; RNA binding motif single stranded interacting protein 2; RNA binding motif, single stranded interacting protein 2; RNA-binding motif, single-stranded-interacting protein 2; SCR3; suppressor of CDC2 with RNA-binding motif 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.