Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBMS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558702
Description
Immunogen sequence: MLLSVTSRPG ISTFGYNRNN KKPYVSLAQQ MAPPSPSNST PNSSSGSNGN DQLSK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 87%.
The protein encoded by this gene is a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. The RBMS proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. This protein was isolated by phenotypic complementation of cdc2 and cdc13 mutants of yeast and is thought to suppress cdc2 and cdc13 mutants through the induction of translation of cdc2.
Specifications
RBMS2 | |
Polyclonal | |
Unconjugated | |
Rbms2 | |
RBMS2; RNA binding motif single stranded interacting protein 2; RNA binding motif, single stranded interacting protein 2; RNA-binding motif, single-stranded-interacting protein 2; SCR3; suppressor of CDC2 with RNA-binding motif 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15434 | |
Rbms2 | |
Recombinant protein corresponding to Human RBMS2. Recombinant protein control fragment (Product #RP-97645). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction