Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566206 Supplier Invitrogen™ Supplier No. PA566206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPTTALQNGFY PAPYNITPNR MLAQSALSPY LSSPVSSYQR VTQTSPLQVP NPSWMHHHSY LMQPSGSVLT PGMDHPISLQ Highest antigen sequence identity to the following orthologs - mouse 57%, rat 83%.

The protein encoded by this gene is a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. The RBMS proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. This protein was isolated by phenotypic complementation of cdc2 and cdc13 mutants of yeast and is thought to suppress cdc2 and cdc13 mutants through the induction of translation of cdc2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMS2
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbms2
Gene Accession No. Q15434
Gene Alias RBMS2; RNA binding motif single stranded interacting protein 2; RNA binding motif, single stranded interacting protein 2; RNA-binding motif, single-stranded-interacting protein 2; SCR3; suppressor of CDC2 with RNA-binding motif 3
Gene Symbols Rbms2
Host Species Rabbit
Immunogen Recombinant Human RBMS2. Recombinant protein control fragment (Product #RP-106890).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.