Learn More
RBMS3 Mouse anti-Human, Clone: OTI8E5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811600

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an RNA-binding protein that belongs to the c-myc gene single-strand binding protein family. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. The encoded protein was isolated by virtue of its binding to an upstream element of the alpha2(I) collagen promoter. The observation that this protein localizes mostly in the cytoplasm suggests that it may be involved in a cytoplasmic function such as controlling RNA metabolism, rather than transcription. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
RBMS3 | |
Monoclonal | |
Unconjugated | |
RBMS3 | |
RNA binding motif single stranded interacting protein 3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27303 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8E5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6XE24 | |
RBMS3 | |
Human recombit protein fragment corresponding to amino acids 1-437 of human RBMS3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.