Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550421 Supplier Invitrogen™ Supplier No. PA550421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total RBMX protein.

hnRNP G is a glycoprotein, which was originally identified as an autoantigen from German shepherd dogs with lupus-like syndrome. The gene encoding hnRNP G is located on chromosome Xq26 and is ubiquitously expressed. It contains one RNP-consensus RNA binding domain (RBD) and is related to RBMY, which is involved in spermatogenesis, and hnRNP G-T, which is a testis specific protein. All three proteins interact with Tra2b and therefore, are involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RBMX
Gene Accession No. P38159
Gene Alias glycoprotein p43; Heterogeneous nuclear ribonucleoprotein G; hnRNP G; HNRNPG; hnRNP-G; Hnrpg; MRXS11; RBMX; Rbmxp1; RBMXRT; RNA binding motif protein, X chromosome; RNA binding motif protein, X-linked; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome retrogene; RNA-binding motif protein, X chromosome retrogene-like; RNA-binding motif protein, X chromosome, N-terminally processed; RNMX
Gene Symbols RBMX
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human RNA binding motif protein, X-linked.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27316
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.