Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBMX Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115371
Description
Antibody detects endogenous levels of total hnRNP G.
hnRNP G is a glycoprotein, which was originally identified as an autoantigen from German shepherd dogs with lupus-like syndrome. The gene encoding hnRNP G is located on chromosome Xq26 and is ubiquitously expressed. It contains one RNP-consensus RNA binding domain (RBD) and is related to RBMY, which is involved in spermatogenesis, and hnRNP G-T, which is a testis specific protein. All three proteins interact with Tra2b and therefore, are involved in pre-mRNA splicing.
Specifications
RBMX | |
Polyclonal | |
Unconjugated | |
RBMX | |
glycoprotein p43; Heterogeneous nuclear ribonucleoprotein G; hnRNP G; HNRNPG; hnRNP-G; Hnrpg; MRXS11; RBMX; Rbmxp1; RBMXRT; RNA binding motif protein, X chromosome; RNA binding motif protein, X-linked; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome retrogene; RNA-binding motif protein, X chromosome retrogene-like; RNA-binding motif protein, X chromosome, N-terminally processed; RNMX | |
Rabbit | |
Affinity chromatography | |
RUO | |
19655, 27316, 302855 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P38159, Q4V898, Q9WV02 | |
RBMX | |
A synthesized peptide derived from human RBMX(Accession P38159), corresponding to amino acid residues N20-N70. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction