Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589752 Supplier Invitrogen™ Supplier No. PA589752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNPLTKVKLI NELNEREVQL GVADKVSWHS EYKDSAWIFL GGLPYELTEG DIICVFSQYG EIVNINLVRD KKTGKSKGFC FLCYEDQRST ILAVDNFNGI KIKGRTIRVD HVSNYRAPKD SEEIDDVTRQ LQEKGCGART PSPSLSESSE DEKPTKKHKK DKKEKKKKKK EKEKADREVQ AEQPSSSSPR RKTVKEKDDT GPKKHSSKNS ERAQKSEPRE GQKLPKSRTA YSGGAEDLER ELKKEKPKHE HKSSSRREAR EEKTRIRDRG RSSDAHSSWY NGRSEGRSYR SRSRSRDKSH RHKRARRSRE RESSNPSDRW RH; Positive Samples: Mouse liver.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMX2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RBMX2
Gene Accession No. Q8R0F5
Gene Alias 2700089A17Rik; 2810411A03Rik; CGI-79; Rbmx2; RGD1562693; RNA binding motif protein, X-linked 2; RNA-binding motif protein, X-linked 2; Snu17
Gene Symbols RBMX2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-322 of human RBMX2 (NP_0571082).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209003
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.