Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMXL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571250 Supplier Invitrogen™ Supplier No. PA571250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 92%; Rabbit: 86%; Rat: 92%; Zebrafish: 86%.

This gene represents a retrogene of RNA binding motif protein, X-linked (RBMX), which is located on chromosome X. While all introns in the coding sequence have been processed out compared to the RBMX locus, the ORF is intact and there is specific evidence for transcription at this location. The preservation of the ORF by purifying selection in all Old World monkeys carrying it suggests that this locus is likely to be functional, possibly during male meiosis when X chromosomal genes are silenced or during haploid stages of spermatogenesis. This gene shares 5' exon structure with the cysteine conjugate-beta lyase 2 locus on chromosome 1, but the coding sequences are non-overlapping. Alternative splicing results in two transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMXL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBMXL1
Gene Accession No. Q96E39
Gene Alias heterogeneous nuclear ribonucleoprotein G (hnRNP G); heterogeneous nuclear ribonucleoprotein G-like 1; hnRNP G; Hnrpg; RBM1; Rbmx; Rbmxl1; Rbmxrt; RNA binding motif protein, X chromosome retrogene; RNA binding motif protein, X linked-like-1; RNA binding motif protein, X-linked like 1; RNA binding motif protein, X-linked-like 1; RNA binding motif protein, X-linked-like-1; RNA-binding motif protein, X chromosome
Gene Symbols RBMXL1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human RBMXL1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494115
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.