Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMXL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541848 Supplier Invitrogen™ Supplier No. PA541848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSRDGYSSRD YREPRGFAPS PGEYTHRDYG HSSVRDDCPL RGYSDRDGYG Sequence homology: Cow: 85%; Dog: 77%; Guinea Pig: 77%; Horse: 77%; Human: 100%; Mouse: 77%; Rabbit: 85%; Rat: 85%.

The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. RBMXL2 has two RRM domains that bind RNAs. RBMXL2 has similarity to HNRPG and RBMY proteins and it is suggested to replace HNRPG protein function during meiotic prophase or act as a germ cell-specific splicing regulator. It primarily localizes to the nuclei of meiotic spermatocytes. This gene is a candidate for autosomal male infertility.This gene belongs to the HNRPG subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMXL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBMXL2
Gene Accession No. O75526
Gene Alias 1700012H05Rik; Heterogeneous nuclear ribonucleoprotein G; heterogeneous nuclear ribonucleoprotein G T; hnRNP G; hnRNP G-T; HNRNPGT; Hnrnpg-t; HNRPG; HNRPGT; LOC617969 protein; NEWGENE_1598286; RBMX; RBMX like 2; Rbmxl2; RNA binding motif protein, X-linked like 2; RNA binding motif protein, X-linked-like 2; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome, N-terminally processed; RNA-binding motif protein, X-linked-like-2; testes specific heterogenous nuclear ribonucleoprotein G T; testes-specific heterogenous nuclear ribonucleoprotein G-T; testis-specific heterogeneous nuclear ribonucleoprotein G-T
Gene Symbols RBMXL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RBMXL2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27288
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.