Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBMXL2 Recombinant Rabbit Monoclonal Antibody (JE64-33)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544704
This item is not returnable.
View return policy
Description
RBMXL2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. RBMXL2 has two RRM domains that bind RNAs. RBMXL2 has similarity to HNRPG and RBMY proteins and it is suggested to replace HNRPG protein function during meiotic prophase or act as a germ cell-specific splicing regulator. It primarily localizes to the nuclei of meiotic spermatocytes. This gene is a candidate for autosomal male infertility.This gene belongs to the HNRPG subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs).
Specifications
RBMXL2 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O75526 | |
RBMXL2 | |
Recombinant protein within human hnRNP G-T aa 151-350/392. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
JE64-33 | |
Unconjugated | |
RBMXL2 | |
1700012H05Rik; Heterogeneous nuclear ribonucleoprotein G; heterogeneous nuclear ribonucleoprotein G T; hnRNP G; hnRNP G-T; HNRNPGT; Hnrnpg-t; HNRPG; HNRPGT; LOC617969 protein; NEWGENE_1598286; RBMX; RBMX like 2; Rbmxl2; RNA binding motif protein, X-linked like 2; RNA binding motif protein, X-linked-like 2; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome, N-terminally processed; RNA-binding motif protein, X-linked-like-2; testes specific heterogenous nuclear ribonucleoprotein G T; testes-specific heterogenous nuclear ribonucleoprotein G-T; testis-specific heterogeneous nuclear ribonucleoprotein G-T | |
Rabbit | |
Protein A | |
RUO | |
103690209, 27288, 76572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction