Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMY1A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112872
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112872 Supplier Invitrogen™ Supplier No. PA5112872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBMY1A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

RBMY1A1 is a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. This protein is thought to function as a splicing regulator during spermatogenesis. Multiple closely related paralogs of this gene are found in a gene cluster in the AZFb azoospermia factor region of chromosome Y. Most of these related copies are thought to be pseudogenes, though several likely encode functional proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMY1A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RBMY1A1
Gene Accession No. P0DJD3
Gene Alias hRBMY; RBM; RBM1; RBM2; RBMY; RBMY1A1; RBMY1C; RNA binding motif protein, Y-linked, family 1, member A1; RNA-binding motif protein 1; RNA-binding motif protein 2; RNA-binding motif protein, Y chromosome, family 1 member A1; RNA-binding motif protein, Y chromosome, family 1 member A1/C; Y chromosome RNA recognition motif 1; Y chromosome RNA recognition motif 2; YRRM1; YRRM2
Gene Symbols RBMY1A1
Host Species Rabbit
Immunogen Recombinant Human RNA-binding motif protein, Y chromosome, family 1 member A1 protein (1-459AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5940
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.