Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBMY1F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562740 Supplier Invitrogen™ Supplier No. PA562740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGSTCHAYSN TRDRYGRSWE SYSSCGDFHY CDREHVCRKD QRNPPSLGRV LPDPREA Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 43%.

This gene encodes a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. Multiple copies of this gene are found in the AZFb azoospermia factor region of chromosome Y and the encoded protein is thought to be involved in spermatogenesis. Most copies of this locus are pseudogenes, although six highly similar copies have full-length ORFs and are considered functional. Four functional copies of this gene are found within inverted repeat IR2; two functional copies of this gene are found in palindrome P3, along with two copies of PTPN13-like, Y-linked.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBMY1F
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBMY1F
Gene Accession No. Q15415
Gene Alias RBMY1F; RBMY1J; RNA binding motif protein, Y-linked, family 1, member F; RNA-binding motif protein, Y chromosome, family 1 member F/J; Y chromosome RNA recognition motif 2; YRRM2
Gene Symbols RBMY1F
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBMY1F. Recombinant protein control fragment (Product #RP-104486).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159163
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.