Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP1 Monoclonal Antibody (G4E4)
GREENER_CHOICE

Catalog No. 01672086
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-086 Supplier Invitrogen™ Supplier No. MA135782
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

G4E4 recognizes an epitope within the 74-182 C-terminal sequence (11kD peptide fragment) of human serum Cellular Retinol Binding Protein 1 (CRBP 1), a single-chain glycoprotein belonging to the superfamily of hydrophobic molecule transporter proteins, which is responsible for transport of retinol (vitamin A1) from the liver to peripheral target tissues, like the eye, where it mediates the cellular uptake. CRBP 1 is synthesized by hepatic parenchymal cells where it becomes bound to its ligand retinol and is then released into the circulation, where it binds further to the protein transthyretin, to form a transporting complex, which is big enough not to be lost by filtration through the kidney glomeruli. It is detected in nearly all tissues with higher expression in adult ovary, pancreas, pituitary gland, adrenal gland, and fetal liver.

This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP1
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone G4E4
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Rbp1
Gene Accession No. P02696, P09455, Q00915
Gene Alias C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; RBP1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellular
Gene Symbols Rbp1
Host Species Mouse
Immunogen Human retinol binding protein (RBP) purified from plasma.
Purification Method purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100354448, 102189996, 19659, 25056, 5947
Target Species Human, Mouse, Rat, Rabbit, Monkey, Goat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.