Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP1 Monoclonal Antibody (OTI2H3)
GREENER_CHOICE

Catalog No. PIMA525518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525518 Supplier Invitrogen™ Supplier No. MA525518
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rbp1
Gene Accession No. P09455
Gene Alias C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; RBP1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellular
Gene Symbols Rbp1
Host Species Mouse
Immunogen Full length human recombinant protein of RBP1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5947
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.