Learn More
RBP1 Mouse anti-Human, Clone: OTI2E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502835

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
RBP1 | |
Monoclonal | |
Unconjugated | |
RBP1 | |
CRABP-I, CRBP, CRBP1, CRBPI, RBPC | |
Mouse | |
Affinity Chromatography | |
RUO | |
5947 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09455 | |
RBP1 | |
Full length human recombit protein of human RBP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.