Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

$178.40 - $482.00

Specifications

Antigen RBP1
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119391
GREENER_CHOICE
View Documents
Invitrogen™
PA5119391
200 μL
Each for $482.00
Only null left
Add to Cart
 
PIPA5121651
View Documents
Invitrogen™
PA5121651
20 μL
Each for $178.40
Only null left
Add to Cart
 
Description

Description

RBP1 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

RBP1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Rat
Rbp1
C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; RBP1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellular
Rbp1
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P02696, P09455
25056, 5947
Recombinant protein Retinol Binding Protein 1, Cellular. The antigen corresponds to amino acid range 2-135 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.