Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP2 Monoclonal Antibody (D2)

Mouse Monoclonal Antibody

$213.50 - $522.50

Specifications

Antigen RBP2
Clone D2
Concentration 1 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA541670
View Documents
Invitrogen™
MA541670
200 μL
Each for $522.50
Only null left
Add to Cart
 
PIMA543614
View Documents
Invitrogen™
MA543614
20 μL
Each for $213.50
Only null left
Add to Cart
 
Description

Description

RBP2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

RBP2 is an abundant protein present in the small intestinal epithelium. It is thought to participate in the uptake and/or intracellular metabolism of vitamin A. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. RBP2 may also modulate the supply of retinoic acid to the nuclei of endometrial cells during the menstrual cycle.
Specifications

Specifications

RBP2
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.05% ProClin 300
P50120
5948
Recombinant protein Retinol Binding Protein 2, Cellular. The antigen corresponds to amino acid range 1-134 of the target protein.
Antibody
D2
-20°C
Monoclonal
Liquid
IgG2b κ
Human
RBP2
AA409370; C76986; Cellular retinol-binding protein II; CRABP-II; CRBP2; Crbp-2; CrbpII; CRBP-II; Histone demethylase JARID1A; Jarid1a; jumonji, AT rich interactive domain 1A (Rbp2 like); Jumonji, AT rich interactive domain 1A (RBP2-like); jumonji/ARID domain-containing protein 1A; KDM5A; lysine (K)-specific demethylase 5A; lysine demethylase 5A; lysine-specific demethylase 5A; RBBP2; RBBP-2; RBP2; Rbp-2; RBPC2; retinoblastoma binding protein 2; retinoblastoma-binding protein 2; retinol binding protein 2; retinol binding protein 2, cellular; retinol-binding protein 2; Retinol-binding protein 2 cellular; retinol-binding protein 2, cellular
RBP2
Primary
Protein A/G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.