Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP2 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145249
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145249 200 μL
FEPA5145250 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145249 Supplier Invitrogen™ Supplier No. PA5145249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBP2 is an abundant protein present in the small intestinal epithelium. It is thought to participate in the uptake and/or intracellular metabolism of vitamin A. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. RBP2 may also modulate the supply of retinoic acid to the nuclei of endometrial cells during the menstrual cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene RBP2
Gene Accession No. P06768, P50120, P50121, Q08652
Gene Alias AA409370; C76986; Cellular retinol-binding protein II; CRABP-II; CRBP2; Crbp-2; CrbpII; CRBP-II; Histone demethylase JARID1A; Jarid1a; jumonji, AT rich interactive domain 1A (Rbp2 like); Jumonji, AT rich interactive domain 1A (RBP2-like); jumonji/ARID domain-containing protein 1A; KDM5A; lysine (K)-specific demethylase 5A; lysine demethylase 5A; lysine-specific demethylase 5A; RBBP2; RBBP-2; RBP2; Rbp-2; RBPC2; retinoblastoma binding protein 2; retinoblastoma-binding protein 2; retinol binding protein 2; retinol binding protein 2, cellular; retinol-binding protein 2; Retinol-binding protein 2 cellular; retinol-binding protein 2, cellular
Gene Symbols RBP2
Host Species Rabbit
Immunogen Recombinant Human RBP2 protein, Met1-Lys134 (Accession #P50120).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19660, 24710, 407611, 5948
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.