Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592951 Supplier Invitrogen™ Supplier No. PA592951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPTHLFQPSL VLDMAKVLLD NYCFPENLLG MQEAIQQAIK SHEILSISDP QTLASVLTAG VQSSLNDPRL VISYEPSTPE PPPQVPALTS LSEEELLAWL QRGLRHEVLE GNVGYLRVDS VPGQEVLSMM GEFLVAHVWG NLMGTSALVL DLRHCTGGQV SGIPYIISYL HPGNTILHVD TIYNRPSNTT TEIWTLPQVL GERYGADKDV VVLTSSQTRG VAEDIAHILK QMRRAIVVGE RTGGGALDLR KLRIGESDFF FTVPVSRSLG PLGGGSQTWE GSGVLPCVGT PAEQALEKAL AIL; Positive Samples: Mouse lung, Mouse eye, Rat eye; Cellular Location: extracellular matrix, extracellular space, interphotoreceptor matrix.

Interphotoreceptor retinol-binding protein is a large glycoprotein known to bind retinoids and found primarily in the interphotoreceptor matrix of the retina between the retinal pigment epithelium and the photoreceptor cells. It is thought to transport retinoids between the retinal pigment epithelium and the photoreceptors, a critical role in the visual process.The human IRBP gene is approximately 9.5 kbp in length and consists of four exons separated by three introns. The introns are 1.6-1.9 kbp long. The gene is transcribed by photoreceptor and retinoblastoma cells into an approximately 4.3-kilobase mRNA that is translated and processed into a glycosylated protein of 135,000 Da. The amino acid sequence of human IRBP can be divided into four contiguous homology domains with 33-38% identity, suggesting a series of gene duplication events. In the gene, the boundaries of these domains are not defined by exon-intron junctions, as might have been expected. The first three homology domains and part of the fourth are all encoded by the first large exon, which is 3,180 base pairs long. The remainder of the fourth domain is encoded in the last three exons, which are 191, 143, and approximately 740 base pairs long, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBP3
Gene Accession No. P10745, P49194
Gene Alias D10S64; D10S65; D10S66; interphotoreceptor retinoid binding protein; interphotoreceptor retinoid-binding protein; interphotoreceptor retinol- binding protein; interstitial retinol binding protein 3; interstitial retinol-binding protein; interstitial retinol-binding protein 3; IRBP; RBP3; Rbp-3; RBPI; retinol binding protein 3; retinol binding protein 3, interstitial; retinol-binding protein 3; RP66
Gene Symbols RBP3
Host Species Rabbit
Immunogen Recombinant protein or (fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19661, 24711, 5949
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.