Learn More
Invitrogen™ RBP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590882
Description
Immunogen sequence: ERDCRVSSFR VKENFDKARF SGTWYAMAKK DPEGLFLQDN IVAEFSVDET GQMSATAKGR VRLLNNWDVC ADMVGTFTDT EDPAKFKMKY WGVASFLQKG NDDHWIVDTD YDTYAVQYSC RLLNLDGTCA DSYSFVFSRD PNGLPPEAQK IVRQRQEELC LARQYRLIVH NGYCDGRSER NLL; Positive Samples: SW620, HeLa, OVCAR3, Mouse liver, Mouse spleen.
This protein belongs to the lipocalin family and is the specific carrier for retinol in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post translationally and results in defective delivery and supply to the epidermal cells.
Specifications
RBP4 | |
Polyclonal | |
Unconjugated | |
RBP4 | |
MCOPCB10; Plasma retinol binding protein 4 (PRBP); Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; PRO2222; RBP; Rbp4; Rbp-4; RBPA; RDCCAS; retinol; retinol binding; retinol binding protein 4; Retinol binding protein 4 interstitial; retinol binding protein 4, cellular; retinol binding protein 4, plasma; retinol-binding protein 4; retinol-binding protein 4, interstitial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19662, 5950 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02753, Q00724 | |
RBP4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 19-201 of human RBP4 (NP_0067352). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.